missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RHOBTB1 Partial ORF (AAH32848, 494 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH32848 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 9886 |
Molekulargewicht | 37.73kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16161416
|
Abnova™
H00009886-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16151416
|
Abnova™
H00009886-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene belongs to the Rho family of the small GTPase superfamily. It contains a GTPase domain, a proline-rich region, a tandem of 2 BTB (broad complex, tramtrack, and bric-a-brac) domains, and a conserved C-terminal region. The protein plays a role in small GTPase-mediated signal transduction and the organization of the actin filament system. Alternate splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: GAISAHKPLLICSCEWMAAMFGGSFVESANSEVYLPNINKISMQAVLDYLYTKQLSPNLDLDPLELIALANRFCLPHLVALAEQHAVQELTKAATSGVGIDGEVLSYLELSpezifikation
AAH32848 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0740/MGC33059/MGC33841 | |
RHOBTB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9886 | |
RHOBTB1 (Human) Recombinant Protein (Q01) | |
GAISAHKPLLICSCEWMAAMFGGSFVESANSEVYLPNINKISMQAVLDYLYTKQLSPNLDLDPLELIALANRFCLPHLVALAEQHAVQELTKAATSGVGIDGEVLSYLEL | |
RUO | |
RHOBTB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |