Learn More
Abnova™ Human RFX4 Full-length ORF (NP_115880.2, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X3, and X5. It has been shown to interact with itself as well as with regulatory factors X2 and X3, but it does not interact with regulatory factor X1. This protein may be a transcriptional repressor rather than a transcriptional activator. Three transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_115880.2 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 5992 |
Molekulargewicht | 99kDa |
Name | RFX4 (Human) Recombinant Protein (P02) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 ug |
Immunogen | MNWAAFGGSEFFIPEGIQIDSRCPLSRNITEWYHYYGIAVKESSQYYDVMYSKKGAAWVSETGKKEVSKQTVAYSPRSKLGTLLPEFPNVKDLNLPASLPEEKVSTFIMMYRTHCQRILDTVIRANFDEVQSFLLHFWQGMPPHMLPVLGSSTVVNIVGVCDSILYKAISGVLMPTVLQALPDSLTQVIRKFAKQLDEWLKVALHDLPENLRNIKFELSRRFSQILRRQTSLNHLCQASRTVIHSADITFQMLEDWRNVDLNSITKQTLYTMEDSRDEHRKLITQLYQEFDHLLEEQSPIESYIEWLDTMVDRCVVKVAAKRQGSLKKVAQQFLLMWSCFGTRVIRDMTLHSAPSFGSFHLIHLMFDDYVLYLLESLHCQERANELMRAMKGEGSTAEVREEIILTEAAAPTPSPVPSFSPAKSATSVEVPPPSSPVSNPSPEYTGLSTTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPHDTAISGPLHYAPYHRSSAQYPFNSPTSRMEPCLMSSTPRLHPTPVTPRWPEVPSANTCYTSPSVHSARYGNSSDMYTPLTTRRNSEYEHMQHFPGFAYINGEASTGWAK |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.