missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RECK Partial ORF (NP_066934, 701 a.a. - 800 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008434-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16138525

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 07-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis. [provided by RefSeq]

Sequence: KFGCSQYECVPRQLACDQVQDPVCDTDHMEHNNLCTLYQRGKSLSYKGPCQPFCRATEPVCGHNGETYSSVCAAYSDRVAVDYYGDCQAVGVLSEHSSVA
Spezifikation

Spezifikation

NP_066934
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KFGCSQYECVPRQLACDQVQDPVCDTDHMEHNNLCTLYQRGKSLSYKGPCQPFCRATEPVCGHNGETYSSVCAAYSDRVAVDYYGDCQAVGVLSEHSSVA
RUO
RECK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8434
RECK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ST15/hRECK
RECK
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote