missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RABGAP1 Full-length ORF (NP_036329.2, 1 a.a. - 997 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00023637-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127582

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MDDQPGEKELVKRSQLDGEGDGPLSNQLSASSTINPVPLVGLQKPEMSLPVKPGQGDSEASSPFTPVADEDSVVFSKLTYLGCASVNAPRSEVEALRMMSILRSQCQISLDVTLSVPNVSEGIVRLLDPQTNTEIANYPIYKILFCVRGHDGTPESDCFAFTESHYNAELFRIHVFRCEIQEAVSRILYSFATAFRRSAKQTPLSATAAPQTPDSDIFTFSVSLEIKEDDGKGYFSAVPKDKDRQCFKLRQGIDKKIVIYVQQTTNKELAIERCFGLLLSPGKDVRNSDMHLLDLESMGKSSDGKSYVITGSWNPKSPHFQVVNEETPKDKVLFMTTAVDLVITEVQEPVRFLLETKVRVCSPNERLFWPFSKRSTTENFFLKLKQIKQRERKNNTDTLYEVVCLESESERERRKTTASPSVRLPQSGSQSSVIPSPPEDDEEEDNDEPLLSGSGDVSKECAEKILETWGELLSKWHLNLNVRPKQLSSLVRNGVPEALRGEVWQLLAGCHNNDHLVEKYRILITKESPQDSAITRDINRTFPAHDYFKDTGGDGQDSLYKICKAYSVYDEEIGYCQGQSFLAAVLLLHMPEEQAFSVLVKIMFDYGLRELFKQNFEDLHCKFYQLERLMQEYIPDLYNHFLDISLEAHMYASQWFLTLFTAKFPLYMVFHIIDLLLCEGISVIFNVALGLLKTSKDDLLLTDFEGALKFFRVQLPKRYRSEENAKKLMELACNMKISQKKLKKYEKEYHTMREQQAQQEDPIERFERENRRLQEANMRLEQENDDLAHELVTSKIALRKDLDNAEEKADALNKELLMTKQKLIDAEEEKRRLEEESAQLKEMCRRELDKAESEIKKNSSIIGDYKQICSQLSERLEKQQTANKVEIEKIRQKVDDCERCREFFNKEGRVKGISSTKEVLDEDTDEEKETLKNQLREMELELAQTKLQLVEAECKIQDLEHHLGLALNEVQAAKKTWFNRTLSSIKTATGVQGKETC
Spezifikation

Spezifikation

NP_036329.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
140.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586D2123/GAPCENA/RP11-123N4.2/TBC1D11
RABGAP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23637
RABGAP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDQPGEKELVKRSQLDGEGDGPLSNQLSASSTINPVPLVGLQKPEMSLPVKPGQGDSEASSPFTPVADEDSVVFSKLTYLGCASVNAPRSEVEALRMMSILRSQCQISLDVTLSVPNVSEGIVRLLDPQTNTEIANYPIYKILFCVRGHDGTPESDCFAFTESHYNAELFRIHVFRCEIQEAVSRILYSFATAFRRSAKQTPLSATAAPQTPDSDIFTFSVSLEIKEDDGKGYFSAVPKDKDRQCFKLRQGIDKKIVIYVQQTTNKELAIERCFGLLLSPGKDVRNSDMHLLDLESMGKSSDGKSYVITGSWNPKSPHFQVVNEETPKDKVLFMTTAVDLVITEVQEPVRFLLETKVRVCSPNERLFWPFSKRSTTENFFLKLKQIKQRERKNNTDTLYEVVCLESESERERRKTTASPSVRLPQSGSQSSVIPSPPEDDEEEDNDEPLLSGSGDVSKECAEKILETWGELLSKWHLNLNVRPKQLSSLVRNGVPEALRGEVWQLLAGCHNNDHLVEKYRILITKESPQDSAITRDINRTFPAHDYFKDTGGDGQDSLYKICKAYSVYDEEIGYCQGQSFLAAVLLLHMPEEQAFSVLVKIMFDYGLRELFKQNFEDLHCKFYQLERLMQEYIPDLYNHFLDISLEAHMYASQWFLTLFTAKFPLYMVFHIIDLLLCEGISVIFNVALGLLKTSKDDLLLTDFEGALKFFRVQLPKRYRSEENAKKLMELACNMKISQKKLKKYEKEYHTMREQQAQQEDPIERFERENRRLQEANMRLEQENDDLAHELVTSKIALRKDLDNAEEKADALNKELLMTKQKLIDAEEEKRRLEEESAQLKEMCRRELDKAESEIKKNSSIIGDYKQICSQLSERLEKQQTANKVEIEKIRQKVDDCERCREFFNKEGRVKGISSTKEVLDEDTDEEKETLKNQLREMELELAQTKLQLVEAECKIQDLEHHLGLALNEVQAAKKTWFNRTLSSIKTATGVQGKETC
RUO
RABGAP1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote