missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB4B Full-length ORF (NP_057238.3, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00053916-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150433

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 31-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

RAB proteins, such as RAB4B, are members of the RAS superfamily of small GTPases that are involved in vesicular trafficking (He et al., 2002 [PubMed 12450215]).[supplied by OMIM]

Sequence: MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGC
Spezifikation

Spezifikation

NP_057238.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ78649/MGC52123
RAB4B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
53916
RAB4B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGC
RUO
RAB4B
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote