missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pyruvate Dehydrogenase Phosphatase/PDP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marke: R&D Systems AF7260
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C Polyclonal specifically detects Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C in Human samples. It is validated for Western Blot.
Spezifikation
Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C | |
Polyclonal | |
Unconjugated | |
Q9P0J1 | |
PDP1 | |
E. coli-derived recombinant human Pyruvate Dehydrogenase Phosphatase/PDP1 Ala72-Glu537 Accession # Q9P0J1 | |
100 μg | |
Primary | |
Detects human Pyruvate Dehydrogenase Phosphatase/PDP1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial, EC 3.1.3, EC 3.1.3.43, MGC119646, PDH, PDP 1, PDP1, PDPC, PDPC 1, PDPFLJ32517, PPM2C, PPM2CFLJ56179, Protein phosphatase 2C, protein phosphatase 2C, magnesium-dependent, catalytic subunit, pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase, Pyruvate dehydrogenase phosphatase catalytic subunit 1, pyruvate dehyrogenase phosphatase catalytic subunit 1 | |
Sheep | |
Affinity Purified | |
RUO | |
54704 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Human Pyruvate Dehydrogenase Phosphatase/PDP1 Antibody, R&D Systems™ > 100μg; Unlabeled
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur