Learn More
R&D Systems™ Human PSCA (NP_005663.2) VersaClone cDNA
Product Code. 18722297
Change view
Click to view available options
Menge:
10 μg
Unit Size:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
18722297 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722297 Supplier R&D Systems™ Supplier No. RDC3336
  • ONLINE EXCLUSIVE PRICE
    326.00 CHF
    307.65 CHF / 10 Mikrogramm
    Sparen 18.35 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Gen PSCA
Zugriffsnummer NP_005663.2
Gen-ID (Entrez) 8000
Gen-Alias PRO232, prostate stem cell antigen
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Human
Menge 10 μg