missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PRSS27 Partial ORF (NP_114154, 94 a.a. - 203 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00083886-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Pancreasin is a pancreatic tryptic serine peptidase that cleaves peptides after an arginine residue.[supplied by OMIM]
Sequence: QLVQPGPHAMYARVRQVESNPLYQGTASSADVALVELEAPVPFTNYILPVCLPDPSVIFETGMNCWVTGWGSPSEEDLLPEPRILQKLAVPIIDTPKCNLLYSKDTEFGYSpezifikation
NP_114154 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QLVQPGPHAMYARVRQVESNPLYQGTASSADVALVELEAPVPFTNYILPVCLPDPSVIFETGMNCWVTGWGSPSEEDLLPEPRILQKLAVPIIDTPKCNLLYSKDTEFGY | |
RUO | |
PRSS27 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
83886 | |
PRSS27 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPH2/MPN | |
PRSS27 | |
Recombinant | |
wheat germ expression system |