Learn More
Abnova™ Human PRPF4 Full-length ORF (NP_004688.2, 1 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009128-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.[supplied by OMIM]
Sequence: MASSRASSTQATKTKAPDDLVAPVVKKPHIYYGSLEEKERERLAKGESGILGKDGLKAGIEAGNINITSGEVFEIEEHISERQAEVLAEFERRKRARQINVSTDDSEVKACLRALGEPITLFGEGPAERRERLRNILSVVGTDALKKTKKDDEKSKKSKEEYQQTWYHEGPNSLKVARLWIANYSLPRAMKRLEEARLHKEIPETTRTSQMQELHKSLRSLNNFCSQIGDDRPISYCHFSPNSKMLATACWSGLCKLWSVPDCNLLHTLRGHNTNVGAIVFHPKSTVSLDPKDVNLASCAADGSVKLWSLDSDEPVADIEGHTVRVARVMWHPSGRFLGTTCYDRSWRLWDLEAQEEILHQEGHSMGVYDIAFHQDGSLAGTGGLDAFGRVWDLRTGRCIMFLEGHLKEIYGINFSPNGYHIATGSGDNTCKVWDLRQRRCVYTIPAHQNLVTGVKFEPIHGNFLLTGAYDNTAKIWTHPGWSPLKTLAGHEGKVMGLDISSDGQLIATCSYDRTFKLWMAESpezifikation
NP_004688.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HPRP4/HPRP4P/PRP4/Prp4p | |
PRPF4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9128 | |
PRPF4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASSRASSTQATKTKAPDDLVAPVVKKPHIYYGSLEEKERERLAKGESGILGKDGLKAGIEAGNINITSGEVFEIEEHISERQAEVLAEFERRKRARQINVSTDDSEVKACLRALGEPITLFGEGPAERRERLRNILSVVGTDALKKTKKDDEKSKKSKEEYQQTWYHEGPNSLKVARLWIANYSLPRAMKRLEEARLHKEIPETTRTSQMQELHKSLRSLNNFCSQIGDDRPISYCHFSPNSKMLATACWSGLCKLWSVPDCNLLHTLRGHNTNVGAIVFHPKSTVSLDPKDVNLASCAADGSVKLWSLDSDEPVADIEGHTVRVARVMWHPSGRFLGTTCYDRSWRLWDLEAQEEILHQEGHSMGVYDIAFHQDGSLAGTGGLDAFGRVWDLRTGRCIMFLEGHLKEIYGINFSPNGYHIATGSGDNTCKVWDLRQRRCVYTIPAHQNLVTGVKFEPIHGNFLLTGAYDNTAKIWTHPGWSPLKTLAGHEGKVMGLDISSDGQLIATCSYDRTFKLWMAE | |
RUO | |
PRPF4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |