missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protein S/PROS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB4036-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Protein S/PROS1 Monoclonal specifically detects Protein S/PROS1 in Human samples. It is validated for Western Blot.
Spezifikation
| Protein S/PROS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P07225 | |
| PROS1 | |
| Mouse myeloma cell line NS0-derived recombinant human Protein S Ala42-Trp670 Accession # P07225 | |
| 25 μg | |
| Primary | |
| Detects human Protein S in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Protein S. | |
| Human | |
| Purified |
| Western Blot | |
| 391609 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, PROS, PROS1, protein S (alpha), protein Sa, PS21, PS22, PS23, PS24, PS25, PSA, vitamin K-dependent plasma protein S, vitamin K-dependent protein S | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5627 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur