missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human PROCR / EPCR / CD201 Protein, Fc Tag

Marke:  ACROBiosystems PRR-H5254-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817557

  • 2072.64 CHF / 1 Milligramm


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Endothelial protein C receptor (PROCR) is also known as Activated protein C receptor (APC receptor), Endothelial cell protein C receptor, CD antigen CD201. PROCR / EPCR / CD201 is a N-glycosylated protein, with a soluble form exists. PROCR is probably released by a metalloprotease. PROCR seems to have the same activity as the membrane-bound form. PROCR / EPCR / CD201 is expressed strongly in the endothelial cells of arteries and veins in heart and lung. PROCR binds activated protein C and enhances protein C activation by the thrombin - thrombomodulin complex. PROCR plays a role in the protein C pathway controlling blood coagulation.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH14451
HEK293
Human
48.6 kDa
1 mg
Ser 18 - Thr 209
PROCR,EPCR,CD201,APC receptor,CCCA,CCD41
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 18 - Thr 209 (Accession # AAH14451).
-20°C
PROCR
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation