Learn More
Novus Biologicals™ Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric)
Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke: Novus Biologicals™ NBP2-76465
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human, Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) measures human Procollagen Type 1 N-Terminal Propeptide in serum, plasma and other biological fluids.
Spezifikation
15,63 bis 1.000 pg/ml | |
ELISA Kit (Colorimetric) | |
The Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) recognizes Human P1NP in samples. No significant cross-reactivity or interference between Human P1NP and analogues was observed. | |
Human | |
Cellular Markers, Extracellular Matrix, Metastasis | |
amino-terminal propeptide, PINP |
9.38 pg/mL | |
Biological Fluids, Plasma, Serum | |
1277 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
Berichtigung von Produktinhalten
Name des Produkts
Novus Biologicals™ Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) > 1 Kit
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur