Learn More
Human Pregnancy Zone Protein/PZP Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  R&D Systems MAB8280-SP
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18784829
  • 259.00 CHF / 25 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Pregnancy Zone Protein Monoclonal specifically detects Pregnancy Zone Protein in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Pregnancy Zone Protein
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6, CPAMD6, pregnancy zone protein, pregnancy-zone protein, PZP
Mouse
Protein A or G purified from cell culture supernatant
RUO
5858
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Western Blot
909813
Western Blot 2 ug/mL
P20742
PZP
Human embryonic kidney cell line HEK293-derived recombinant human Pregnancy Zone Protein/PZP Thr26-Val1482 Accession # P20742
25 μg
Primary
Detects human Pregnancy Zone Protein/PZP in direct ELISAs.
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Aktionen
Aktionen