missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPP3CA Partial ORF (NP_000935, 1 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005530-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MSEPKAIDPKLSTTDRVVKAVPFPPSHRLTAKEVFDNDGKPRVDILKAHLMKEGRLEESVALRIITEGASILRQEKNLLDIDAPSpezifikation
NP_000935 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSEPKAIDPKLSTTDRVVKAVPFPPSHRLTAKEVFDNDGKPRVDILKAHLMKEGRLEESVALRIITEGASILRQEKNLLDIDAP | |
RUO | |
PPP3CA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5530 | |
PPP3CA (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CALN/CALNA/CALNA1/CCN1/CNA1/PPP2B | |
PPP3CA | |
Recombinant | |
wheat germ expression system |