missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PNOC Partial ORF (NP_006219.1, 96 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005368-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: RRMPRVRSLFQEQEEPEPGMEEAGEMEQKQLQKRFGGFTGARKSARKLANQKRFSEFMRQYLVLSMQSSQRRRSpezifikation
NP_006219.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RRMPRVRSLFQEQEEPEPGMEEAGEMEQKQLQKRFGGFTGARKSARKLANQKRFSEFMRQYLVLSMQSSQRRR | |
RUO | |
PNOC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5368 | |
PNOC (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPNOC | |
PNOC | |
Recombinant | |
wheat germ expression system |