missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PLEKHK1 Full-length ORF (AAH25765, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00219790-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MEGPSLRGPALRLAGLPTQQDCNIQEKIDLEIRMREGIWKLLSLSTQKDQVLHAVKNLMVCNARLMAYTSELQKLEEQIANQTGRCDVKFESKERTACKGKIAISDIRIPLMWKDSDHFSNKERSRRYAIFCLFKMGANVFDTDVVNVDKTITDICFENVTILSpezifikation
AAH25765 | |
Liquid | |
219790 | |
PLEKHK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGPSLRGPALRLAGLPTQQDCNIQEKIDLEIRMREGIWKLLSLSTQKDQVLHAVKNLMVCNARLMAYTSELQKLEEQIANQTGRCDVKFESKERTACKGKIAISDIRIPLMWKDSDHFSNKERSRRYAIFCLFKMGANVFDTDVVNVDKTITDICFENVTIL | |
RUO | |
RTKN2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.67kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J10120/PLEKHK1/bA531F24.1 | |
RTKN2 | |
Yes | |
wheat germ expression system |