Learn More
ACROBiosystems Human PLAU / uPA Protein, His Tag
Marke: ACROBiosystems PLU-H5229-50UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Urokinase - type plasminogen activator is also known as PLAU and UPA, a serine protease with an extremely limited substrate specificity, cleaving the sequence Cys – Pro – Gly - Arg560 - Val561 – Val – Gly – Gly – Cys in plasminogen to form plasmin. uPA is a potent marker of invasion and metastasis in a variety of human cancers associated with breast, stomach, colon, bladder, ovary, brain and endometrium.uPA and its receptor (uPAR) have been implicated in a broad spectrum of pathophysiological processes, including fibrinolysis, proteolysis, inflammation, atherogenesis and plaque destabilization, all of which are involved in the pathogenesis of MI (myocardial infarction).
Human PLAU, His Tag is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 47.2 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Please note that the protein is not a
Spezifikation
NP_002649.1 | |
50 μg | |
Ser 21 - Leu 431 | |
Urokinase,PLAU,ATF,UPA,URK,u-PA,BDPLT5,QPD | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
47.2 kDa | |
Human PLAU, His Tag (PLU-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession # NP_002649.1). | |
-20°C | |
PLAU | |
Human | |
50 mM Sodium Acetate, 100 mM NaCl, pH5.0 | |
HEK293 | |
Enzymes |