missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PITPNC1 Full-length ORF (AAH07905, 1 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH07905 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 26207 |
Molekulargewicht | 55.22kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16128172
|
Abnova™
H00026207-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16118172
|
Abnova™
H00026207-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the phosphatidylinositol transfer protein family. The encoded cytoplasmic protein transfers phosphatidylinositol from one membrane compartment to another. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MLLKEYRICMPLTVDEYKIGQLYMISKHSHEQSDRGEGVEVVQNEPFEDPHHGNGQFTEKRVYLNSKLPSWARAVVPKIFYVTEKAWNYYPYTITEYTCSFLPRFSIHIETKYEDNKGSNDTIFDNEAKDVEREVCFIDIACDEIPERYYKESEDPKHFKSEKTGRGQLREGWRDSHQPIMCSYKLVTVKFEVWGLQTRVEQFVHKVVRDILLIGHRQAFAWVDEWYDMTMDDVREYEKNMHEQTNIKVCNQHSSPVDDIESHAQTSTSpezifikation
AAH07905 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.22kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLLKEYRICMPLTVDEYKIGQLYMISKHSHEQSDRGEGVEVVQNEPFEDPHHGNGQFTEKRVYLNSKLPSWARAVVPKIFYVTEKAWNYYPYTITEYTCSFLPRFSIHIETKYEDNKGSNDTIFDNEAKDVEREVCFIDIACDEIPERYYKESEDPKHFKSEKTGRGQLREGWRDSHQPIMCSYKLVTVKFEVWGLQTRVEQFVHKVVRDILLIGHRQAFAWVDEWYDMTMDDVREYEKNMHEQTNIKVCNQHSSPVDDIESHAQTST | |
RUO | |
PITPNC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26207 | |
PITPNC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RDGB-BETA/RDGBB/RDGBB1 | |
PITPNC1 | |
Recombinant | |
wheat germ expression system |