missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human PCSK9 (R194A) Protein, His Tag
auf Anfrage
Spezifikation
Zugriffsnummer | AAI166619.1 |
---|---|
Molekulargewicht | 13.8 kDa and 58.0 kDa |
Quelle | Human PCSK9 (R194A), His Tag (PCA-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # AAI166619.1). |
Immunogen | Gln 31 - Gln 692 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16427949
|
ACROBiosystems
PCA-H5226-1MG |
1 mg |
auf Anfrage
1 Milligramm |
N/A | |||||
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann. Fordern Sie hier einen Preis für diesen Artikel an. |
|||||||||
15817447
|
ACROBiosystems
PCA-H5226-50UG |
50 μg |
auf Anfrage
50 Mikrogramm |
N/A | |||||
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann. Fordern Sie hier einen Preis für diesen Artikel an. |
|||||||||
Beschreibung
Proprotein convertase subtilisin/kexin type 9 (PCSK9), is an enzyme which in humans is encoded by the PCSK9 gene.This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein plays a major regulatory role in cholesterol homeostasis. PCSK9 binds to the epidermal growth factor-like repeat A (EGF-A) domain of the low-density lipoprotein receptor (LDLR), inducing LDLR degradation. PCSK9 may also have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a rare form of autosomal dominant familial hypercholesterolemia (HCHOLA3).
This protein is fused with a polyhistidine tag at the C-terminus. This protein undergoes autocatalytic cleavage to release the pro-peptide and mature chain. The pro-peptide and mature chain are associated through non‐covalent interactions and with a calcuSpezifikation
AAI166619.1 | |
Human PCSK9 (R194A), His Tag (PCA-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # AAI166619.1). | |
-20°C | |
PCSK9 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
13.8 kDa and 58.0 kDa | |
Gln 31 - Gln 692 | |
PCSK9,FH3,HCHOLA3,LDLCQ1,NARC1,PC9 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |