Learn More
Abnova™ Human PAX6 Full-length ORF (NP_000271.1, 1 a.a. - 422 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_000271.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 5080 |
Molekulargewicht | 73.1kDa |
Name | PAX6 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 ug |
Immunogen | MQNSHSGVNQLGGVFVNGRPLPDSTRQKIVELAHSGARPCDISRILQVSNGCVSKILGRYYETGSIRPRAIGGSKPRVATPEVVSKIAQYKRECPSIFAWEIRDRLLSEGVCTNDNIPSVSSINRVLRNLASEKQQMGADGMYDKLRMLNGQTGSWGTRPGWYPGTSVPGQPTQDGCQQQEGGGENTNSISSNGEDSDEAQMRLQLKRKLQRNRTSFTQEQIEALEKEFERTHYPDVFARERLAAKIDLPEARIQVWFSNRRAKWRREEKLRNQRRQASNTPSHIPISSSFSTSVYQPIPQPTTPVSSFTSGSMLGRTDTALTNTYSALPPMPSFTMANNLPMQPPVPSQTSSYSCMLPTSPSVNGRSYDTYTPPHMQTHMNSQPMGTSGTTSTGLISPGVSVPVQVPGSEPDMSQYWPRLQ |
Mehr anzeigen |
Produktvorschläge
Customers who viewed this item also viewed.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.