missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Park7/DJ-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
292.00 CHF - 569.00 CHF
Spezifikation
| Antigen | Park7/DJ-1 |
|---|---|
| Klon | 925812R |
| Verdünnung | ELISA |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18739198
![]() |
R&D Systems
MAB39952-SP |
25 μg |
292.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18719208
![]() |
R&D Systems
MAB39952-100 |
100 μg |
569.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human samples. It is validated for ELISA.Spezifikation
| Park7/DJ-1 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q99497 | |
| 11315 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 925812R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| E. coli-derived recombinant human/mouse/rat Park7/DJ-1 Ala2-Asp189 Accession # Q99497 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Park7/DJ-1 in direct ELISAs. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts