Learn More
ACROBiosystems Human OX40 / TNFRSF4 / CD134 Protein, His Tag
MALS verified
Marke: ACROBiosystems OX0-H5224-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003318.1 | |
1 mg | |
Leu 29 - Ala 216 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
22.0 kDa | |
Human O x 40 Protein, His Tag (MALS verified) (O x 0-H5224) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # P43489-1). | |
-20°C | |
OX40 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |