missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL5 Partial ORF (NP_663613, 4 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_663613 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 114879 |
Molekulargewicht | 36.3kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16131437
|
Abnova™
H00114879-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 11-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16121437
|
Abnova™
H00114879-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 11-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: EAFLRRRFSLCPPSSTPQKVDPRKLTRNLLLSGDNELYPLSPGKDMEPNGPSLPRDEGPPTPSSATKVPPAEYRLCNGSDKECVSPTARVTKKETLSpezifikation
NP_663613 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42929/OBPH1/ORP5 | |
OSBPL5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
114879 | |
OSBPL5 (Human) Recombinant Protein (Q01) | |
EAFLRRRFSLCPPSSTPQKVDPRKLTRNLLLSGDNELYPLSPGKDMEPNGPSLPRDEGPPTPSSATKVPPAEYRLCNGSDKECVSPTARVTKKETL | |
RUO | |
OSBPL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |