missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Orexin A ELISA Kit
For the quantitative detection of Human Orexin A
Marke: Invitrogen™ EEL039
Additional Details : Gewicht : 0.40000kg
Beschreibung
This gene encodes a hypothalamic neuropeptide precursor protein that gives rise to two mature neuropeptides, orexin A and orexin B, by proteolytic processing. Orexin A and orexin B, which bind to orphan G-protein coupled receptors HCRTR1 and HCRTR2, function in the regulation of sleep and arousal. This neuropeptide arrangement may also play a role in feeding behavior, metabolism, and homeostasis.Spezifikation
O43612 | |
37.5 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
HCRT | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
62.5 to 4000 pg/mL | |
HRP | |
Plasma | |
3060 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
50 µL | |
Human | |
3 hr 30 min |