Learn More
Abnova™ Human OR2C3 Partial ORF (NP_932340.2, 220 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spécification
Zugriffsnummer | NP_932340.2 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 81472 |
Molekulargewicht | 36.63kDa |
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
16140387
|
Abnova™
H00081472-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16130387
|
Abnova™
H00081472-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]
Sequence: HIARAVLKIRSAEGRRKAFNTCSSHVAVVSLFYGSIIFMYLQPAKSTSHEQGKFIALFYTVVTPALNPLIYTLRNTEVKSALRHMVLENCCGSAGKLAQISpécification
NP_932340.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OR2C4/OR2C5P/OST742 | |
OR2C3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
81472 | |
OR2C3 (Human) Recombinant Protein (Q01) | |
HIARAVLKIRSAEGRRKAFNTCSSHVAVVSLFYGSIIFMYLQPAKSTSHEQGKFIALFYTVVTPALNPLIYTLRNTEVKSALRHMVLENCCGSAGKLAQI | |
RUO | |
OR2C3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |