Learn More
Abnova™ Human NUP54 Full-length ORF (NP_059122.2, 1 a.a. - 507 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly (FG) repeat-containing nucleoporin subset. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_059122.2 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 53371 |
Molekulargewicht | 81.8kDa |
Name | NUP54 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | MAFNFGAPSGTSGTAAATAAPAGGFGGFGTTSTTAGSAFSFSAPTNTGTTGLFGGTQNKGFGFGTGFGTTTGTSTGLGTGLGTGLGFGGFNTQQQQQTTLGGLFSQPTQAPTQSNQLINTASALSAPTLLGDERDAILAKWNQLQAFWGTGKGYFNNNIPPVEFTQENPFCRFKAVGYSCMPSNKDEDGLVVLVFNKKETEIRSQQQQLVESLHKVLGGNQTLTVNVEGTKTLPDDQTEVVIYVVERSPNGTSRRVPATTLYAHFEQANIKTQLQQLGVTLSMTRTELSPAQIKQLLQNPPAGVDPIIWEQAKVDNPDSEKLIPVPMVGFKELLRRLKVQDQMTKQHQTRLDIISEDISELQKNQTTSVAKIAQYKRKLMDLSHRTLQVLIKQEIQRKSGYAIQADEEQLRVQLDTIQGELNAPTQFKGRLNELMSQIRMQNHFGAVRSEERYYIDADLLREIKQHLKQQQEGLSHLISIIKDDLEDIKLVEHGLNETIHIRGGVFS |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.