Learn More
Abnova™ Human NFS1 Partial ORF (NP_858931.1, 29 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_858931.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 9054 |
Molekulargewicht | 37.84kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16129825
|
Abnova™
H00009054-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16119825
|
Abnova™
H00009054-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Iron-sulfur clusters are required for the function of many cellular enzymes. The protein encoded by this gene supplies inorganic sulfur to these clusters by removing the sulfur from cysteine, creating alanine in the process. This gene uses alternate in-frame translation initiation sites to generate mitochondrial forms and cytoplasmic/nuclear forms. Selection of the alternative initiation sites is determined by the cytosolic pH. The encoded protein belongs to the class-V family of pyridoxal phosphate-dependent aminotransferases. [provided by RefSeq]
Sequence: LRLRVGDRAPQSAVPADTAAAPEVGPVLRPLYMDVQATTPLDPRVLDAMLPYLINYYGNPHSRTHAYGWESEAAMERARQQVASLIGADPREIIFTSGATESNNIAIKTESpezifikation
NP_858931.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUSSY-08/IscS/NIFS | |
NFS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9054 | |
NFS1 (Human) Recombinant Protein (Q01) | |
LRLRVGDRAPQSAVPADTAAAPEVGPVLRPLYMDVQATTPLDPRVLDAMLPYLINYYGNPHSRTHAYGWESEAAMERARQQVASLIGADPREIIFTSGATESNNIAIKTE | |
RUO | |
NFS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |