missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NENF Full-length ORF (NP_037481.1, 1 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00029937-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129212

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a neurotrophic factor that may play a role in neuron differentiation and development. A pseudogene of this gene is found on chromosome 12. Alternate splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Sequence: MVGPAPRRRLRPLAALALVLALAPGLPTARAGQTPRPAERGPPVRLFTEEELARYGGEEEDQPIYLAVKGVVFDVTSGKEFYGRGAPYNALTGKDSTRGVAKMSLDPADLTHDTTGLTAKELEALDEVFTKVYKAKYPIVGYTARRILNEDGSPNLDFKPEDQPHFDIKDEF
Spezifikation

Spezifikation

NP_037481.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIR2/NEUDESIN/SCIRP10/SPUF
NENF
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29937
NENF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVGPAPRRRLRPLAALALVLALAPGLPTARAGQTPRPAERGPPVRLFTEEELARYGGEEEDQPIYLAVKGVVFDVTSGKEFYGRGAPYNALTGKDSTRGVAKMSLDPADLTHDTTGLTAKELEALDEVFTKVYKAKYPIVGYTARRILNEDGSPNLDFKPEDQPHFDIKDEF
RUO
NENF
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote