missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEDD9 Full-length ORF (NP_006394.1, 1 a.a. - 834 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_006394.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 4739 |
Molekulargewicht | 119.3kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16130668
|
Abnova™
H00004739-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16120668
|
Abnova™
H00004739-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Sequence: MKYKNLMARALYDNVPECAEELAFRKGDILTVIEQNTGGLEGWWLCSLHGRQGIVPGNRVKLLIGPMQETASSHEQPASGLMQQTFGQQKLYQVPNPQAAPRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPVFSVPVGEIKPQGVYDIPPTKGVYAIPPSACRDEAGLREKDYDFPPPMRQAGRPDLRPEGVYDIPPTCTKPAGKDLHVKYNCDIPGAAEPVARRHQSLSPNHPPPQLGQSVGSQNDAYDVPRGVQFLEPPAETSEKANPQERDGVYDVPLHNPPDAKGSRDLVDGINRLSFSSTGSTRSNMSTSSTSSKESSLSASPAQDKRLFLDPDTAIERLQRLQQALEMGVSSLMALVTTDWRCYGYMERHINEIRTAVDKVELFLKEYLHFVKGAVANAACLPELILHNKMKRELQRVEDSHQILSQTSHDLNECSWSLNILAINKPQNKCDDLDRFVMVAKTVPDDAKQLTTTINTNAEALFRPGPGSLHLKNGPESIMNSTEYPHGGSQGQLLHPGDHKAQAHNKALPPGLSKEQAPDCSSSDGSERSWMDDYDYVHLQGKEEFERQQKELLEKENIMKQNKMQLEHHQLSQFQLLEQEITKPVENDISKWKPSQSLPTTNSGVSAQDRQLLCFYYDQCETHFISLLNAIDALFSCVSSAQPPRIFVAHSKFVILSAHKLVFIGDTLTRQVTAQDIRNKVMNSSNQLCEQLKTIVMATKMAALHYPSTTALQEMVHQVTDLSRNAQLFKRSLLEMATFSpezifikation
NP_006394.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKYKNLMARALYDNVPECAEELAFRKGDILTVIEQNTGGLEGWWLCSLHGRQGIVPGNRVKLLIGPMQETASSHEQPASGLMQQTFGQQKLYQVPNPQAAPRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPVFSVPVGEIKPQGVYDIPPTKGVYAIPPSACRDEAGLREKDYDFPPPMRQAGRPDLRPEGVYDIPPTCTKPAGKDLHVKYNCDIPGAAEPVARRHQSLSPNHPPPQLGQSVGSQNDAYDVPRGVQFLEPPAETSEKANPQERDGVYDVPLHNPPDAKGSRDLVDGINRLSFSSTGSTRSNMSTSSTSSKESSLSASPAQDKRLFLDPDTAIERLQRLQQALEMGVSSLMALVTTDWRCYGYMERHINEIRTAVDKVELFLKEYLHFVKGAVANAACLPELILHNKMKRELQRVEDSHQILSQTSHDLNECSWSLNILAINKPQNKCDDLDRFVMVAKTVPDDAKQLTTTINTNAEALFRPGPGSLHLKNGPESIMNSTEYPHGGSQGQLLHPGDHKAQAHNKALPPGLSKEQAPDCSSSDGSERSWMDDYDYVHLQGKEEFERQQKELLEKENIMKQNKMQLEHHQLSQFQLLEQEITKPVENDISKWKPSQSLPTTNSGVSAQDRQLLCFYYDQCETHFISLLNAIDALFSCVSSAQPPRIFVAHSKFVILSAHKLVFIGDTLTRQVTAQDIRNKVMNSSNQLCEQLKTIVMATKMAALHYPSTTALQEMVHQVTDLSRNAQLFKRSLLEMATF | |
RUO | |
NEDD9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4739 | |
NEDD9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAS-L/CAS2/CASL/CASS2/HEF1/dJ49G10.2/dJ761I2.1 | |
NEDD9 | |
Recombinant | |
wheat germ expression system |