Learn More
ACROBiosystems Human Nectin-3 / CD113 Protein, His Tag
378.31 CHF - 2522.00 CHF
Spezifikation
Zugriffsnummer | Q9NQS3 |
---|---|
Molekulargewicht | 39.6 kDa |
Quelle | Human Nectin-3, His Tag (PV3-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3). |
Immunogen | Gly 58 - Asp 400 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817577
|
ACROBiosystems
PV3-H52E4-100UG |
100 μg |
378.31 CHF
100 Mikrogramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827577
|
ACROBiosystems
PV3-H52E4-1MG |
1 mg |
2522.00 CHF
1 Milligramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Poliovirus receptor-related 3 (PVRL3), also known as nectin-3 and CD113, is a human protein of the immunoglobulin superfamily which forms part of adherens junctions. Nectins are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin system in epithelial cells. PVRL3 plays a role in cell-cell adhesion through heterophilic trans-interactions with nectin-like proteins or nectins, such as trans-interaction with PVRL2/nectin-2 at Sertoli-spermatid junctions. Furthermore, PVRL3 induces endocytosis-mediated down-regulation of PVR from the cell surface, resulting in reduction of cell movement and proliferation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.6 kDa. The protein migrates as 55-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
Q9NQS3 | |
Human Nectin-3, His Tag (PV3-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3). | |
-20°C | |
Nectin-3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
39.6 kDa | |
Gly 58 - Asp 400 | |
CD113,Nectin-3,PVRL3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |