missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDST3 Partial ORF (NP_004775, 162 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00009348-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150436

  • 295.00 CHF / 10 Mikrogramm


Verfügbar ab
11-08-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: IGFHKTSEKSVQSFQLKGFPFSIYGNLAVKDCCINPHSPLIRVTKSSKLEKGSLPGTDWTVFQINHSAYQPVIFAKVKTPENLSPSISKGAFYATIIH
Spezifikation

Spezifikation

NP_004775
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IGFHKTSEKSVQSFQLKGFPFSIYGNLAVKDCCINPHSPLIRVTKSSKLEKGSLPGTDWTVFQINHSAYQPVIFAKVKTPENLSPSISKGAFYATIIH
RUO
NDST3
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9348
NDST3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSST3|MGC130028|MGC130029
NDST3
Yes
Videos
Dokumentation