missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDST3 Partial ORF (NP_004775, 162 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

auf Anfrage

Spezifikation

Name NDST3 (Human) Recombinant Protein (Q01)
Rekombinant Yes
Molekulargewicht 36.52kDa
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

This gene encodes a member of the heparan sulfate/heparin GlcNAc N-deacetylase/ N-sulfotransferase family. The encoded enzyme is a type II transmembrane protein that resides in the Golgi apparatus. This monomeric bifunctional enzyme catalyzes the N-deacetylation and N-sulfation of N-acetylglucosamine residues in heparan sulfate and heparin, which are the initial chemical modifications required for the biosynthesis of the functional oligosaccharide sequences that define the specific ligand binding activities of heparan sulfate and heparin. [provided by RefSeq]

Sequence: IGFHKTSEKSVQSFQLKGFPFSIYGNLAVKDCCINPHSPLIRVTKSSKLEKGSLPGTDWTVFQINHSAYQPVIFAKVKTPENLSPSISKGAFYATIIH
Spezifikation

Spezifikation

NDST3 (Human) Recombinant Protein (Q01)
Yes
36.52kDa
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications