missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NCAM1 Partial ORF (AAH47244, 611 a.a. - 710 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004684-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: EPSAPKLEGQMGEDGNSIKVNLIKQDDGGSPIRHYLVRYRALSSEWKPEIRLPSGSDHVMLKSLDWNAEYEVYVVAENQQGKSKAAHFVFRTSAQPTAIPSpezifikation
AAH47244 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPSAPKLEGQMGEDGNSIKVNLIKQDDGGSPIRHYLVRYRALSSEWKPEIRLPSGSDHVMLKSLDWNAEYEVYVVAENQQGKSKAAHFVFRTSAQPTAIP | |
RUO | |
NCAM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4684 | |
NCAM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD56/MSK39/NCAM | |
NCAM1 | |
Recombinant | |
wheat germ expression system |