missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NAV2 Full-length ORF (AAH16054.1, 1 a.a. - 873 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00089797-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126393

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene.[supplied by OMIM]

Sequence: MSLSNPTMLRTHSLSNADGQYDPYTDSRFRNSSMSLDEKSRTMSRSGSFRDGFEEVHGSSLSLVSSTSSVYSTPEEKCQSEIRKLRRELDASQEKVSALTTQLTANAHLVAAFEQSLGNMTIRLQSLTMTAEQKDSELNELRKTIELLKKQNAAAQAAINGVINTPELNCKGNGTAQSADLRIRRQHSSDSVSSINSATSHSSVGSNIESDSKKKKRKNWLRSSFKQAFGKKSPKSASSHSDIEEMTDSSLPSSPKLPHNGSTGSTPLLRNSHSNSLISECMDSEAETVMQLRNELRDKEMKLTDIRLEALSSAHQLDQLREAMNRMQSEIEKLKAENDRLKSESQGSGCSRAPSQVSISASPRQSMGLSQHSLNLTESTSLDMLLDDTGECSARKEGGRHVKIVVSFQEEMKWKEDSRPHLFLIGCIGVSGKTKWDVLDGVVRRLFKEYIIHVDPVSQLGLNSDSVLGYSIGEIKRSNTSETPELLPCGYLVGENTTISVTVKGLAENSLDSLVFESLIPKPILQRYVSLLIEHRRIILSGPSGTGKTYLANRLSEYIVLREGRELTDGVIATFNVDHKSSKELRQYLSNLADQCNSENNAVDMPLVIILDNLHHVSSLGEIFNGLLNCKYHKCPYIIGTMNQATSSTPNLQLHHNFRWVLCANHTEPVKGFLGRFLRRKLMETEISGRVRNMELVKIIDWIPKVWHHLNRFLEAHSSSDVTIGPRLFLSCPIDVDGSRVWFTDLWNYSIIPYLLEAVREGLQLYGRRAPWEDPAKWVMDTYPWAASPQQHEWPPLLQLRPEDVGFDGYSMPREGSTSKQMPPSDAEGDPLMNMLMRLQEAANYSSPQSYDSDSNSNSHHDDILDSSLESTL
Spezifikation

Spezifikation

AAH16054.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
124kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10633/FLJ11030/FLJ23707/FLJ77876/HELAD1/KIAA1419/POMFIL2/RAINB1/STEERIN2/UNC53H2
NAV2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
89797
NAV2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLSNPTMLRTHSLSNADGQYDPYTDSRFRNSSMSLDEKSRTMSRSGSFRDGFEEVHGSSLSLVSSTSSVYSTPEEKCQSEIRKLRRELDASQEKVSALTTQLTANAHLVAAFEQSLGNMTIRLQSLTMTAEQKDSELNELRKTIELLKKQNAAAQAAINGVINTPELNCKGNGTAQSADLRIRRQHSSDSVSSINSATSHSSVGSNIESDSKKKKRKNWLRSSFKQAFGKKSPKSASSHSDIEEMTDSSLPSSPKLPHNGSTGSTPLLRNSHSNSLISECMDSEAETVMQLRNELRDKEMKLTDIRLEALSSAHQLDQLREAMNRMQSEIEKLKAENDRLKSESQGSGCSRAPSQVSISASPRQSMGLSQHSLNLTESTSLDMLLDDTGECSARKEGGRHVKIVVSFQEEMKWKEDSRPHLFLIGCIGVSGKTKWDVLDGVVRRLFKEYIIHVDPVSQLGLNSDSVLGYSIGEIKRSNTSETPELLPCGYLVGENTTISVTVKGLAENSLDSLVFESLIPKPILQRYVSLLIEHRRIILSGPSGTGKTYLANRLSEYIVLREGRELTDGVIATFNVDHKSSKELRQYLSNLADQCNSENNAVDMPLVIILDNLHHVSSLGEIFNGLLNCKYHKCPYIIGTMNQATSSTPNLQLHHNFRWVLCANHTEPVKGFLGRFLRRKLMETEISGRVRNMELVKIIDWIPKVWHHLNRFLEAHSSSDVTIGPRLFLSCPIDVDGSRVWFTDLWNYSIIPYLLEAVREGLQLYGRRAPWEDPAKWVMDTYPWAASPQQHEWPPLLQLRPEDVGFDGYSMPREGSTSKQMPPSDAEGDPLMNMLMRLQEAANYSSPQSYDSDSNSNSHHDDILDSSLESTL
RUO
NAV2
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote