missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MYO1F Partial ORF (NP_036467.2, 811 a.a. - 912 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004542-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: KKKVDIQALRGVSLSTRQDDFFILQEDAADSFLESVFKTEFVSLLCKRFEEATRRPLPLTFSDTLQFRVKKEGWGGGGTRSVTFSRGFGDLAVLKVGGRTLTSpezifikation
NP_036467.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKKVDIQALRGVSLSTRQDDFFILQEDAADSFLESVFKTEFVSLLCKRFEEATRRPLPLTFSDTLQFRVKKEGWGGGGTRSVTFSRGFGDLAVLKVGGRTLT | |
RUO | |
MYO1F | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4542 | |
MYO1F (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MYO1F | |
Wheat Germ (in vitro) | |
GST | |
Liquid |