missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYLK Full-length ORF (NP_444259.1, 1 a.a. - 153 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00004638-P02.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16130488

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 30-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene, a muscle member of the immunoglobulin gene superfamily, encodes myosin light chain kinase which is a calcium/calmodulin dependent enzyme. This kinase phosphorylates myosin regulatory light chains to facilitate myosin interaction with actin filaments to produce contractile activity. This gene encodes both smooth muscle and nonmuscle isoforms. In addition, using a separate promoter in an intron in the 3' region, it encodes telokin, a small protein identical in sequence to the C-terminus of myosin light chain kinase, that is independently expressed in smooth muscle and functions to stabilize unphosphorylated myosin filaments. A pseudogene is located on the p arm of chromosome 3. Four transcript variants that produce four isoforms of the calcium/calmodulin dependent enzyme have been identified as well as two transcripts that produce two isoforms of telokin. Additional variants have been identified but lack full length transcripts. [provided by RefSeq]

Sequence: MAMISGLSGRKSSTGSPTSPLNAEKLESEDVSQAFLEAVAEEKPHVKPYFSKTIRDLEVVEGSAARFDCKIEGYPDPEVVWFKDDQSIRESRHFQIDYDEDGNCSLIISDVCGDDDAKYTCKAVNSLGEATCTAELIVETMEEGEGEGEEEEE
Spezifikation

Spezifikation

NP_444259.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686I10125/FLJ12216/KRP/MLCK/MLCK1/MLCK108/MLCK210/MSTP083/MYLK1/smMLCK
MYLK
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4638
MYLK (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMISGLSGRKSSTGSPTSPLNAEKLESEDVSQAFLEAVAEEKPHVKPYFSKTIRDLEVVEGSAARFDCKIEGYPDPEVVWFKDDQSIRESRHFQIDYDEDGNCSLIISDVCGDDDAKYTCKAVNSLGEATCTAELIVETMEEGEGEGEEEEE
RUO
MYLK
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote