missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MVP ELISA Kit
For the quantitative detection of Human MVP
Marke: Invitrogen™ EEL044
Additional Details : Gewicht : 0.40000kg
Beschreibung
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.Spezifikation
Q14764 | |
0.47 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
MVP | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
0.78 to 50 ng/mL | |
HRP | |
Plasma | |
9961 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
100 µL | |
Human | |
3 hr 30 min |