missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MTMR9 Partial ORF (NP_056273, 450 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00066036-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129856

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate. [provided by RefSeq]

Sequence: LQQKTMSLWSWVNQPSELSKFTNPLFEANNLVIWPSVAPQSLPLWEGIFLRWNRSSKYLDEAYEEMVNIIEYNKELQAKVNILRRQLAELETEDGMQESP
Spezifikation

Spezifikation

NP_056273
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LQQKTMSLWSWVNQPSELSKFTNPLFEANNLVIWPSVAPQSLPLWEGIFLRWNRSSKYLDEAYEEMVNIIEYNKELQAKVNILRRQLAELETEDGMQESP
RUO
MTMR9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
66036
MTMR9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C8orf9/DKFZp434K171/LIP-STYX/MGC126672/MTMR8
MTMR9
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote