missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Cyclophilin A Antibody, R&D Systems™

Goat Polyclonal Antibody
Marke: R&D Systems AF3589-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cyclophilin A Polyclonal specifically detects Cyclophilin A in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Spezifikation
| Cyclophilin A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cyclophilin A, Cyclosporin A-binding protein, CYPA, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIA, PPIase A, Rotamase, Rotamase A, T cell cyclophilin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5478 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P62937 | |
| PPIA | |
| E. coli-derived recombinant human Cyclophilin A Met1-Glu165 Accession # P62937 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Cyclophilin A in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur