missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse IL-12 R beta 2 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
365.00 CHF - 666.00 CHF
Spezifikation
| Antigen | IL-12 R beta 2 |
|---|---|
| Klon | 305719 |
| Verdünnung | Flow Cytometry 10 uL/10^6 cells |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18790439
![]() |
R&D Systems
FAB1959C-025 |
25 Tests |
365.00 CHF
25 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18788137
![]() |
R&D Systems
FAB1959C-100 |
100 Tests |
666.00 CHF
100 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Flow Cytometry.Spezifikation
| IL-12 R beta 2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| IL12RB2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human and mouse IL-12 R beta 2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-1 R beta 1 is observed. |
| 305719 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| Human, Mouse | |
| Q99665 | |
| 3595 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts