missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD44 Rat, APC-Cy7, Clone: IM7, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB39-APC7-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Human/Mouse CD44 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1, pH7.2 | |
Cd44 | |
Protein A/G Purified | |
RUO | |
12505 | |
IgG2b κ |
Flow Cytometry | |
IM7 | |
APC-Cyanine7 | |
NP_001034239 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |