missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Caspase-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Marke: R&D Systems AF-605-NA
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Knockout Validated.
Spezifikation
| Caspase-3 | |
| Polyclonal | |
| Unconjugated | |
| AAA65015 | |
| CASP3 | |
| E. coli-derived recombinant human Caspase-3 Met1-His277 Accession # AAA65015 | |
| 100 μg | |
| Primary | |
| Detects human Caspase-3. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 1 ug/10^6 cells, Knockout Validated | |
| Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 836 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur