missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Caspase-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Marke: R&D Systems AF-605-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Knockout Validated.
Spezifikation
| Caspase-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 836 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 1 ug/10^6 cells, Knockout Validated | |
| AAA65015 | |
| CASP3 | |
| E. coli-derived recombinant human Caspase-3 Met1-His277 Accession # AAA65015 | |
| 25 μg | |
| Primary | |
| Detects human Caspase-3. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction