Learn More
Abnova™ Human MMP19 Full-length ORF (NP_002420.1, 1 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004327-P02.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This protein is expressed in human epidermis and it has a role in cellular proliferation as well as migration and adhesion to type I collagen. Multiple transcript variants encoding distict isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MNCQQLWLGFLLPMTVSGRVLGLAEVAPVDYLSQYGYLQKPLEGSNNFKPEDITEALRAFQEASELPVSGQLDDATRARMRQPRCGLEDPFNQKTLKYLLLGRWRKKHLTFRILNLPSTLPPHTARAALRQAFQDWSNVAPLTFQEVQAGAADIRLSFHGRQSSYCSNTFDGPGRVLAHADIPELGSVHFDEDEFWTEGTYRGVNLRIIAAHEVGHALGLGHSRYSQALMAPVYEGYRPHFKLHPDDVAGIQALYGKKSPVIRDEEEEETELPTVPPVPTEPSPMPDPCSSELDAMMLGPRGKTYAFKGDYVWTVSDSGPGPLFRVSALWEGLPGNLDAAVYSPRTQWIHFFKGDKVWRYINFKMSPGFPKKLNRVEPNLDAALYWPLNQKVFLFKGSGYWQWDELARTDFSSYPKPIKGLFTGVPNQPSAAMSWQDGRVYFFKGKVYWRLNQQLRVEKGYPRNISHNWMHCRPRTIDTTPSGGNTTPSGTGITLDTTLSATETTFEYSpezifikation
NP_002420.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MMP18/RASI-1 | |
MMP19 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4327 | |
MMP19 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNCQQLWLGFLLPMTVSGRVLGLAEVAPVDYLSQYGYLQKPLEGSNNFKPEDITEALRAFQEASELPVSGQLDDATRARMRQPRCGLEDPFNQKTLKYLLLGRWRKKHLTFRILNLPSTLPPHTARAALRQAFQDWSNVAPLTFQEVQAGAADIRLSFHGRQSSYCSNTFDGPGRVLAHADIPELGSVHFDEDEFWTEGTYRGVNLRIIAAHEVGHALGLGHSRYSQALMAPVYEGYRPHFKLHPDDVAGIQALYGKKSPVIRDEEEEETELPTVPPVPTEPSPMPDPCSSELDAMMLGPRGKTYAFKGDYVWTVSDSGPGPLFRVSALWEGLPGNLDAAVYSPRTQWIHFFKGDKVWRYINFKMSPGFPKKLNRVEPNLDAALYWPLNQKVFLFKGSGYWQWDELARTDFSSYPKPIKGLFTGVPNQPSAAMSWQDGRVYFFKGKVYWRLNQQLRVEKGYPRNISHNWMHCRPRTIDTTPSGGNTTPSGTGITLDTTLSATETTFEY | |
RUO | |
MMP19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |