missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human MMP-2 Recombinant Protein, PeproTech®
Click to view available options
Menge:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
2 Stück
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
420-02-1MG will be provided as 2 x 500 μg (420-02-500UG). Recombinant Human MMP-2 is a 62.0 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain (552 amino acids). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.
Spezifikation
Spezifikation
| Zugriffsnummer | P08253 |
| Zur Verwendung mit (Anwendung) | Functional Assay |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 4313 |
| Molekulargewicht | 62 kDa |
| Name | Human MMP-2 |
| Menge | 100 μg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur