missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human MICA Protein, His Tag

Marke:  ACROBiosystems MIA-H5221-100UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827307

  • 287.87 CHF / 100 Mikrogramm


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

MHC class I polypeptide-related sequence A (MICA) belongs to the MHC class I family and MIC subfamily. MICA contains one Ig-like C1-type (immunoglobulin-like) domain. Unlike classical MHC class I molecules, MICA does not form a heterodimer with beta-2-microglobulin. MICA acts as a stress-induced self-antigen that is recognized by gamma delta T-cells. MICA is ligand for the KLRK1/NKG2D receptor. MICA bind to KLRK1 leads to cell lysis.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.8 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH16929.1
HEK293
Human
33.8 kDa
100 µg
Glu 24 - Gln 308
MIC-A
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Other Recombinant Proteins
Powder
>95% as determined by SDS-PAGE
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # AAH16929.1).
-20°C
MICA
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation