Learn More
ACROBiosystems Human MICA Protein, His Tag
Marke: ACROBiosystems MIA-H5221-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
MHC class I polypeptide-related sequence A (MICA) belongs to the MHC class I family and MIC subfamily. MICA contains one Ig-like C1-type (immunoglobulin-like) domain. Unlike classical MHC class I molecules, MICA does not form a heterodimer with beta-2-microglobulin. MICA acts as a stress-induced self-antigen that is recognized by gamma delta T-cells. MICA is ligand for the KLRK1/NKG2D receptor. MICA bind to KLRK1 leads to cell lysis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.8 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH16929.1 | |
1 mg | |
Glu 24 - Gln 308 | |
MIC-A | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
33.8 kDa | |
Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # AAH16929.1). | |
-20°C | |
MICA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |