missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Mesothelin / MSLN (296-580) Protein, Fc Tag

MALS verified

Marke:  ACROBiosystems MSN-H526X-100UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827327

  • 322.42 CHF / 100 Mikrogramm


Verfügbar ab
26-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.

This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 59.1 kDa. The protein migrates as 60-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH09272
HEK293
Human
59.1 kDa
100 µg
Glu 296 - Gly 580
MSLN,Mesothelin,MPF
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human Mesothelin (296-580), Fc Tag (MSN-H526x) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272).
-20°C
Mesothelin
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation