missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melanopsin Antibody, R&D Systems™

Mouse Monoclonal Antibody
239.00 CHF - 522.00 CHF
Spezifikation
| Antigen | Melanopsin |
|---|---|
| Klon | 628737 |
| Verdünnung | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Anwendungen | Western Blot, Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18763997
![]() |
R&D Systems
MAB10497-SP |
25 μg |
239.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18764407
![]() |
R&D Systems
MAB10497-100 |
100 μg |
522.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Spezifikation
| Melanopsin | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4 | |
| NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 628737 | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UHM6 | |
| 94233 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts