Learn More
R&D Systems™ Human MEGF11 (NP_115821.2) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | MEGF11 |
| Gen-ID (Entrez) | 84465 |
| Gen-Alias | DKFZp434L121, KIAA1781, multiple EGF-like domains protein 11, multiple EGF-like-domains 11, multiple epidermal growth factor-like domains protein 11 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Produktlinie | R&D Systems |
| Menge | 10 μg |